missing translation for 'onlineSavingsMsg'
Learn More

Poly(3-hexylthiophene-2,5-diyl), regiorandom, Thermo Scientific Chemicals

Catalog No. p-7080005
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45906MC 100 mg
AA45906ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA45906MC Supplier Thermo Scientific Chemicals Supplier No. 045906MC
Only null left

Poly(3-hexylthiophene-2,5-diyl) optimized for use in organic photovoltaic's (OPV) research and devices. It is also used in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104934-50-1
Molecular Formula (C10H14S)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00217686
InChI Key DUFPJSOXRHVDOV-UHFFFAOYSA-N
Synonym poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity
PubChem CID 22950249
IUPAC Name 3-hexyl-2,5-dimethylthiophene
SMILES CCCCCCC1=C(-*)SC(-*)=C1

Specifications

Color Brown
Physical Form Powder/Chunks
Solubility Information Soluble (50mg/ml) in chlorinated solvents such as chloroform,chlorobenzene,dichlorobenzene and trichlorobenzene.
Quantity 100 mg
Sensitivity Light sensitive
Chemical Name or Material Poly (3-hexylthiophene-2,5-diyl)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only