missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(2-vinylpyridine), Average Mw 37,500 (Typical), Average Mn 35,000 (Typical), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 25014-15-7 |
| Physical Form | Neat |
| MDL Number | MFCD00134117 |
| Type | Analytical Standard |
| Quantity | 1 g |
| Grade | Analytical Standard |