missing translation for 'onlineSavingsMsg'
Learn More

Poly(2-ethyl-2-oxazoline), M.W. 500,000

Catalog No. AA4239822
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4239822 100 g
AA4239836 500 g
AA42398A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4239822 Supplier Thermo Scientific Chemicals Supplier No. 04239822

CAS: 25805-17-8 | (C5H9NO)n | 99.13 g/mol

Poly(2-ethyl-2-oxazoline) is used as an adhesion promoter in coatings, in heat sealing and remoisten able hot-melt adhesive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25805-17-8
Molecular Formula (C5H9NO)n
Molecular Weight (g/mol) 99.13
MDL Number MFCD00134377
InChI Key NYEZZYQZRQDLEH-UHFFFAOYSA-N
Synonym 2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
PubChem CID 66412
IUPAC Name 2-ethyl-4,5-dihydro-1,3-oxazole
SMILES CCC(=O)N(-*)CC-*

Specifications

Physical Form Off-white solid
Solubility Information Soluble in water and organic solvents.
Quantity 100 g
Chemical Name or Material Poly (2-ethyl-2-oxazoline), M.W. 500,000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only