Learn More
Poly(2-ethyl-2-oxazoline), M.W. 50,000
CAS: 25805-17-8 | (C5H9NO)n | 99.13 g/mol
Supplier: Thermo Scientific Chemicals 04239636
Description
Poly(2-ethyl-2-oxazoline) is used as an adhesion promoter in coatings, in heat sealing and remoisten able hot-melt adhesive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Poly (2-ethyl-2-oxazoline) | |
| Off-white solid | |
| (C5H9NO)n | |
| 2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline | |
| 500 g | |
| CCC(=O)N(-*)CC-* | |
| 99.13 |
| M.W. 50,000 | |
| 25805-17-8 | |
| MFCD00134377 | |
| Soluble in water and organic solvents. | |
| NYEZZYQZRQDLEH-UHFFFAOYSA-N | |
| 2-ethyl-4,5-dihydro-1,3-oxazole | |
| 66412 |
Chemical Identifiers
| 25805-17-8 | |
| 99.13 | |
| NYEZZYQZRQDLEH-UHFFFAOYSA-N | |
| 66412 | |
| CCC(=O)N(-*)CC-* |
| (C5H9NO)n | |
| MFCD00134377 | |
| 2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline | |
| 2-ethyl-4,5-dihydro-1,3-oxazole |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only