missing translation for 'onlineSavingsMsg'
Learn More

Poly(2-ethyl-2-oxazoline), M.W. 50,000

Catalog No. aa4239622
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Catalog No. Quantity
AA4239622 100 g
AA4239636 500 g
AA42396A4 2.5 kg
3 options

Catalog No. AA4239622

Supplier: Thermo Scientific Chemicals 04239622

Only null left

CAS: 25805-17-8 | (C5H9NO)n | 99.13 g/mol

Poly(2-ethyl-2-oxazoline) is used as an adhesion promoter in coatings, in heat sealing and remoisten able hot-melt adhesive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25805-17-8
Molecular Formula (C5H9NO)n
Molecular Weight (g/mol) 99.13
MDL Number MFCD00134377
InChI Key NYEZZYQZRQDLEH-UHFFFAOYSA-N
Synonym 2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
PubChem CID 66412
IUPAC Name 2-ethyl-4,5-dihydro-1,3-oxazole
SMILES CCC(=O)N(-*)CC-*

Specifications

Color White
Physical Form Off-white solid
Solubility Information Soluble in water and organic solvents.
Quantity 100 g
Chemical Name or Material Poly (2-ethyl-2-oxazoline), M.W. 50,000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only