missing translation for 'onlineSavingsMsg'
Learn More

Poly(2-ethyl-2-oxazoline), M.W. 200,000

CAS: 25805-17-8 | (C5H9NO)n | 99.13 g/mol

Supplier:  Thermo Scientific Chemicals 04239736

 View more versions of this product

Catalog No. AA4239736


Only null left
Explore available promotions

Description

Description

Poly(2-ethyl-2-oxazoline) shows the application in biocompatibility, avoiding accumulation in tissue, and rapid clearance from the bloodstream. It is also adapted to many different functions for both protein and small drug conjugation. It also acts as a perfect substitute for poly(vinyl alcohol) and poly(vinyl pyrrolidone). It is also used as an adhesion promoter in coatings and heat sealing. It behaves as a hydrophilic block and utilized in the synthesis of amphiphilic diblock copolymers by reacting with poly(1,3-trimethylene carbonate) (PTMC) as a hydrophobic block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Poly (2-ethyl-2-oxazoline)
Off-white solid
(C5H9NO)n
2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
500 g
CCC(=O)N(-*)CC-*
99.13
M.W. 200,000
25805-17-8
MFCD00134377
Soluble in water and organic solvents.
NYEZZYQZRQDLEH-UHFFFAOYSA-N
2-ethyl-4,5-dihydro-1,3-oxazole
66412

Chemical Identifiers

25805-17-8
99.13
NYEZZYQZRQDLEH-UHFFFAOYSA-N
66412
CCC(=O)N(-*)CC-*
(C5H9NO)n
MFCD00134377
2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
2-ethyl-4,5-dihydro-1,3-oxazole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only