Learn More
SignalChem Biotech Poly (1:1 dI, dC) Acid 25ug
Catalog No. 502568142
Encompass
The synthetic poly (deoxyinosinic-deoxycytidylic) acid sodium salt double-stranded polymer (synthesized as 1:1 ratio) is used as a substrate for DNA methyltransferases.
Catalog No. 502568142 Supplier SignalChem Biotech Supplier No. P625825
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The synthetic poly (deoxyinosinic-deoxycytidylic) acid sodium salt double-stranded polymer (synthesized as 1:1 ratio) is used as a substrate for DNA methyltransferases.