missing translation for 'onlineSavingsMsg'
Learn More

Poltex Flex Neck Biohazard Shield with Magnetic Base

Catalog No. 22000208
Encompass
Change view
Click to view available options
Length (English):
8.5 in.
12 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22000208 8.5 in.
22000209 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22000208 Supplier Poltex Supplier No. 8.5FLEXBIOM
Get a Poltex Pipette Holder with $1,500 Purchase
View all promotions

Flexible neck of 12 in. holds firmly in whatever position per user requirements

  • Made from durable scratch and break-resistant polycarbonate
  • Available in two sizes - 8.5 x 12 in. and 12 x 12 in.

Specifications

Dimensions (L x W) 8.5 x 12 in.
Width (English) 12 in.
Product Type Flexible Neck Biohazard Shield
Includes Magnetic base
Length (English) 8.5 in.
Material Polycarbonate