missing translation for 'onlineSavingsMsg'
Learn More

Poltex Ergonomic Biohazard Shield

Catalog No. 22000183
Encompass
Change view
Click to view available options
Height (English):
11 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
22000183 11 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22000183 Supplier Poltex Supplier No. BIOSH
Get a Poltex Pipette Holder with $1,500 Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Helps to protect the user from biohazardous fluids

  • Made from durable 0.25 in. thick PETG
  • Ergonomic cut outs for each arm allow a more neutral posture
  • Base and face are flared out wider for better splash protection
  • Non-slip rubber bumpons on base
  • Easily disassembles for storage and cleaning
  • Dimensions: 12 inch wide (10 inch at cut out)
  • Base is 10 inches deep
  • Angled shield is 11 inch tall
  • Total height is 16 in.

Specifications

Height (English) 11 in.
Height (Metric) 28 cm
Width (Metric) 30.4 cm
Width (English) 12 in.
Product Type Ergonomic Biohazard Shield