Learn More
Poltex Articulating Biohazard Shield
Catalog No. 22000184
Encompass
Change view
Click to view available options
Product Type:
One articulated stiff stem
Two articulated stiff stems
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22000184 One articulated stiff stem
22000185 Two articulated stiff stems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22000184 Supplier Poltex Supplier No. ARTSHLD7
Provides full range of motion to protect from biohazard splash
  • Constructed of break resistant polycarbonate (3/16 base, 1/8 shield)
  • Comes in two versions: 6.75 in.× 6.75 in. with one stem, and 11.75 in.×11.75 in. with two stems
  • Articulating stem allows maximum range of motion to protect the user from biohazard splash
  • Shield and base units are 6.75 in.× 6.75 in. or 11.75 in.×11.75 in.
  • Rubber bumpons add a non-slip base, and a stiff stem holds the shield in place
Specifications
Product Type One articulated stiff stem
Material Polycarbonate