missing translation for 'onlineSavingsMsg'
Learn More

Poloxamer 407, NF, Spectrum™ Chemical

Catalog No. p-7158010
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
25 kg
45 kg
Packaging:
Fiber Drum
Poly Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605284 2.5 kg Poly Pail
18605286 12 kg Poly Pail
18605287 25 kg Fiber Drum
18605285 45 kg Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605284 Supplier Spectrum Chemical Mfg Cor Supplier No. P11662.5KG
Only null left

P1166, 9003-11-6, HO(C2H4O)a(C3H6O)b(C2H4O)aH

Spectrum™ Chemical Poloxamer 407, NF is a national formulary approved surfactant that finds utility because of its amphiphilic structure. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9003-11-6
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
InChI Key RVGRUAULSDPKGF-UHFFFAOYNA-N
IUPAC Name 2-methyloxirane; oxirane
SMILES C1CO1.CC1CO1

Specifications

pH 5.0 to 7.5
CAS Max % 1
Quantity 2.5 kg
Grade NF
Packaging Poly Pail