missing translation for 'onlineSavingsMsg'
Learn More

Poloxamer 188, NF, Spectrum™ Chemical

Catalog No. p-7158482
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605273 500 g Poly Bottle
18605275 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18605273 Supplier Spectrum Chemical Mfg Cor Supplier No. P1169500GM
Only null left

P1169, 9003-11-6, HO(C2H4O)a(C3H6O)b(C2H4O)aH

Spectrum™ Chemical Poloxamer 188, NF is a useful surfactant for the formulation of cosmetics and pharmaceuticals because it increases miscibility. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9003-11-6
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
InChI Key RVGRUAULSDPKGF-UHFFFAOYNA-N
IUPAC Name 2-methyloxirane; oxirane
SMILES C1CO1.CC1CO1

Specifications

pH 5.0 to 7.5
CAS Max % 1
Quantity 500 g
Grade NF
Packaging Poly Bottle