missing translation for 'onlineSavingsMsg'
Learn More

Poloxamer 184, Surfactant, Spectrum™ Chemical

Catalog No. p-7158007
Change view
Click to view available options
Quantity:
2.5 L
20 L
55 gal.
Packaging:
Drum
Glass Bottle
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605270 2.5 L Glass Bottle
18605271 20 L Steel Can
18605272 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605270 Supplier Spectrum Chemical Mfg Cor Supplier No. P11632.5LTGL
Only null left

P1163, 9003-11-6, HO(C2H4O)a(C3H6O)b(C2H4O)aH

Spectrum™ Chemical Poloxamer 184, Surfactant is a nonionic triblock copolymer. It is made up of a main hydrophobic chain of polyoxypropylene bordered on each side by two hydrophilic chains of polyoxyethylene. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 9003-11-6
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
InChI Key RVGRUAULSDPKGF-UHFFFAOYNA-N
IUPAC Name 2-methyloxirane; oxirane
SMILES C1CO1.CC1CO1

Specifications

pH 5.0 to 7.5
CAS Max % 1
Quantity 2.5 L
Grade Reagent
Packaging Glass Bottle
Water 0.004