missing translation for 'onlineSavingsMsg'
Learn More

Poloxamer 124, Spectrum™ Chemical

Catalog No. 18605259
Change view
Click to view available options
Quantity:
20 L
200 L
Packaging:
Drum
Poly Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605259 20 L Poly Drum
18605260 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18605259 Supplier Spectrum Chemical Mfg Cor Supplier No. P116820LT

P1168, 9003-11-6, HO(C2H4O)a(C3H6O)b(C2H4O)aH

Spectrum™ Chemical Poloxamer 124 has an amphiphilic structure (both hydrophilic and lipophilic) and is a useful surfactant for the formulation of cosmetics and pharmaceuticals because it increases miscibility. Poloxamer 124 has also been used to control thickness (viscosity) of personal care products and dish soaps.

Chemical Identifiers

CAS 9003-11-6
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
InChI Key RVGRUAULSDPKGF-UHFFFAOYNA-N
IUPAC Name 2-methyloxirane; oxirane
SMILES C1CO1.CC1CO1

Specifications

pH 5.0 to 7.5
CAS Max % 1
Quantity 20 L
Grade Reagent
Packaging Poly Drum
Water 0.004