missing translation for 'onlineSavingsMsg'
Learn More

Poloxamer 124, Spectrum™ Chemical

Catalog No. 18605258
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Amber Glass Bottle
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605258 2.5 L Glass Bottle
18605257 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18605258 Supplier Spectrum Chemical Mfg Cor Supplier No. P11682.5LTGL
Only null left

P1168, 9003-11-6, HO(C2H4O)a(C3H6O)b(C2H4O)aH

Spectrum™ Chemical Poloxamer 124 has an amphiphilic structure (both hydrophilic and lipophilic) and is a useful surfactant for the formulation of cosmetics and pharmaceuticals because it increases miscibility. Poloxamer 124 has also been used to control thickness (viscosity) of personal care products and dish soaps.

Chemical Identifiers

CAS 9003-11-6
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
InChI Key RVGRUAULSDPKGF-UHFFFAOYNA-N
IUPAC Name 2-methyloxirane; oxirane
SMILES C1CO1.CC1CO1

Specifications

pH 5.0 to 7.5
CAS Max % 1
Quantity 2.5 L
Grade Reagent
Packaging Glass Bottle
Water 0.004