missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Polishers for Milli-Q™ IQ 7000 Ultrapure Water System

Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.

$613.00 - $1,400.46

Specifications

Description Delivered hermetically sealed
For Use With (Equipment) Milli-Q IQ 7000 Ultrapure Water System
Purity Meter Included No
View More Specs

Products 3
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
LCPAK00A1
View Documents Promotion Details
MilliporeSigma™
LCPAK00A1
LC-Pak™ Polisher
Each for $1,400.46
Only null left
 
CDUFBI0A1
View Documents Promotion Details
MilliporeSigma™
CDUFBI0A1
Biopak™ Polisher
Each for $613.00
Only null left
 
EDSPAK0A1
View Documents Promotion Details
MilliporeSigma™
EDSPAK0A1
EDS-Pak™ Polisher
Each for $818.00
Only null left
 
Description

Description

Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.

For use with Milli-Q™ IQ 7000 Ultrapure Water System.
Specifications

Specifications

Delivered hermetically sealed
Milli-Q IQ 7000 Ultrapure Water System
No
Videos
SDS
Documents

Documents