missing translation for 'onlineSavingsMsg'
Learn More

StatLab™ PolarStat Plus Embedding Medium

Catalog No. p-200020511
Change view
Click to view available options
Color:
Blue
Green
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
STLPSP12B Blue
STLPSP12G Green
STLPSP12R Red
STLPSP12Y Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLPSP12B Supplier StatLab™ Supplier No. PSP12B
Only null left
Harmful if swallowed, in contact with skin, or inhaled.

The formulation of this embedding medium freezes at optimum cutting temperatures of -10°C and below.

  • Frozen section embedding medium is a proprietary formulation of water soluble glycols and resins
  • Provides an optimal specimen matrix for Mohs cryostat sectioning, and bonding the specimen securely to the specimen chuck
  • No residue is left on slides during the staining procedure, thereby eliminating undesirable background staining
  • Comes in convenient 4 oz. squeeze bottle

Specifications

Color Blue
Product Type Embedding Media
Quantity 12 Bottles/Cs.
For Use With (Application) Tissue Embedding
Form Liquid