missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StatLab™ PolarStat Plus Embedding Medium
Description
- Frozen section embedding medium is a proprietary formulation of water soluble glycols and resins
- Provides an optimal specimen matrix for Mohs cryostat sectioning, and bonding the specimen securely to the specimen chuck
- No residue is left on slides during the staining procedure, thereby eliminating undesirable background staining
- Comes in convenient 4 oz. squeeze bottle
Specifications
Specifications
| Color | Yellow |
| Product Type | Embedding Media |
| Quantity | 12 Bottles/Cs. |
| For Use With (Application) | Tissue Embedding |
| Form | Liquid |