Thomas Scientific Polarseal Foil Adhesive Tape, for Multiwell Plates, 100 Sheets per Pack
Catalog No. NC9934048
Ideal for long term compound storage Effective temperature range from -80 C to +150 C DMSO resistant Pierceable with a pipet tip for easy sample retrieval Peels off easily to reduce backspray
Catalog No.
NC9934048
Supplier
Thomas ScientificSupplier No.
1152A34
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ideal for long term compound storage Effective temperature range from -80 C to +150 C DMSO resistant Pierceable with a pipet tip for easy sample retrieval Peels off easily to reduce backspray