missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PolarScreen™ ER Alpha Competitor Assay, Green

Catalog No. A15883
Encompass
Change view
Click to view available options
Quantity:
2500 Assays
800 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15883 800 Assays
A15882 2500 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15883 Supplier Thermo Scientific™ Supplier No. A15883

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The PolarScreen™ ER (Estrogen Receptor) Alpha Competitor Assay Kit, Green, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential ER alpha ligands.

  • The PolarScreen™ ER (Estrogen Receptor) Alpha Competitor Assay Kit, Green, provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential ER alpha ligands. The kit uses insect cell-expressed, full-length, native (untagged) human estrogen receptor alpha (available separately, Cat. No. A15674) and a novel, high-affinity, fluorescent estrogen ligand (Fluormone ES2 Green) in a homogenous, mix-and-read assay format.
  • This kit produces a green color read-out; a red kit is also available. Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation.
  • Complete & ready-to-use out of the box - all reagents included, just add your test compounds
  • High throughput compatible - 800 x 20 μL fluorescence polarization assays in a 384-well plate
  • Convenient - available in bulk sizes for larger screening needs
  • Includes:
    • 2 tubes full-length ER Alpha Recombinant Protein (store at -68 to -85°C)
    • 1 tube of Fluormone ES2 Green Tracer (store at -5 to -30°C)
    • 1 bottle of ES2 Screening Buffer (store at -5 to -30°C)

Specifications

Content And Storage Contains:

• 2 tubes full-length ER Alpha Recombinant Protein (store at -68 to -85°C)
• 1 tube of Fluormone ES2 Green Tracer (store at -5 to -30°C)
• 1 bottle of ES2 Screening Buffer (store at -5 to -30°C)
Detection Method Fluorescence
For Use With (Equipment) Fluorescent Imager
Format 384-well Plate
Quantity 800 Assays
Type ER Alpha Competitor Assay
I'm using the TaqMan hPSC Scorecard Panel. Do I need to use the template files?

We highly recommend that you use the template file because it contains all of the necessary experimental details for your qPCR run. If the template file is not working for you, please send an email to our stem cell technical support specialists at: stemcell-help@thermofisher.com.

What are the common issues encountered with FP assays?

-No assay window: Check that your instrument is designed to do FP experiments.
-Poor discrimination between the minimum and maximum mP Control (both include the receptor): This may be an indication of the quality of the receptor. Do not make single use aliquots of the receptor or store diluted.
-The type of microplate is critical: Please see if the protocol specifies untreated polystyrene or NBS-coated plates. If the plate binds the fluorescent substrate, it cannot freely rotate and the mP reading will always be high. White plates cannot be used for FP assays. Avoid the use of silanized pipette tips and vials.
-The “No Receptor Control' (free Fluormone/Tracer) should have much higher RFU values compared to buffer alone. If not, there is a problem with the instrument setup or less commonly with the Fluormone/Tracer.
-If the free Fluormone/Tracer mP value is greater than 100 mP or negative, please refer to the user manual for your instrument and reset the G factor.

What is the best type of microplate reader for fluorescence polarization (FP) assays?

For FP assays, plate readers must specifically have FP built in. Please see our instrument compatibility portal (http://www.thermofisher.com/us/en/home/industrial/pharma-biopharma/drug-discovery-development/dd-misc/instrument-compatibility-portal.html).

How can I test the fluorescence polarization (FP) settings of my microplate reader before doing the assay?

We offer a FP One-Step Reference Kit (https://www.thermofisher.com/order/catalog/product/P3088) to detect in green (Ex/Em = 488/535 nm) and red (Ex/Em = 525/590 nm) emission ranges, suitable for the PolarScreen Green and Red Assays. The alternative is to perform the Controls in the respective assay kit prior to performing the actual assay using test compounds.

For Research Use Only. Not for use in diagnostic procedures.