missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Essentials Strip- and Dot-Style Label Dispenser, Blue Acrylic

Catalog No. 03391355
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
03391355 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03391355 Supplier Cole-Parmer™ Essentials Supplier No. 0439691

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Holds and organizes up to six different labels

  • Load labels in their boxes from the rear of dispenser, feed roll strip through the opening
  • Straight, blunt dispensing edge makes tearing off labels clean and easy-no need for scissors
  • Compatible with all PolarSafe labels on rolls
  • Hold three wide boxes, six thin boxes, or a mixture of the two!
Made from durable, easy-to-clean, acrylic plastic, the dispenser has been designed to conveniently dispense labels for your cryogenic vials, tubes or plates. The dispenser has four rubber feet to hold it in place when used on countertops. Unit can also be wall mounted (screws not included).

Order Info

For PolarSafe Dots, see cat. no. 03391187; for PolarSafe Strips, see cat. no. 03391100.

Specifications

Adhesive Type Peel and Stick
Color Blue
Finish Matte
Format Label
Material Polyester
Printable Sides Front
Print Technology Laser
Product Type Label Dispenser
Sterility Non-sterile
For Use With (Application) Laser printer, can even be used in incubators
Min. Operating Temperature (Metric) -196°C
Quantity 1 Ea.
Temperature Range (English) -320.8°F
Dimensions (L x W) 12 x 6 in.
Length (English) 12 in.
Show More Show Less