missing translation for 'onlineSavingsMsg'
Learn More

StatLab Polarchucks

Catalog No. 22000073
Change view
Click to view available options
Product Type:
Polarchuck Circular Leica
Polarchuck Waffle Leica
Polarchuck Waffle Microm
Length (Metric):
30 mm
40 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Length (Metric)
22000073 Polarchuck Circular Leica 40 mm
22000068 Polarchuck Waffle Microm 30 mm
22000069 Polarchuck Waffle Microm 40 mm
22000070 Polarchuck Waffle Leica 30 mm
22000071 Polarchuck Waffle Leica 40 mm
22000072 Polarchuck Circular Leica 30 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22000073 Supplier StatLab Supplier No. POLARCHKL/40
Only null left

PolarChuck cryostat embedding chucks are made of anodized aluminum

  • For Avantik/Microm Cryostats, Waffle Grid, 30 mm
  • Deep rectangular waffle grid or circular pattern creates a more stable and secure tissue mount

Specifications

Product Type Polarchuck Circular Leica
For Use With (Equipment) Cryostat
Disposable Reusable
Length (Metric) 40 mm