Learn More
Solstice Polar Silica-Based SPE Columns, Cyanopropyl (CN), 500 mg/3 mL
Catalog No. 6002871
Change view
Click to view available options
Quantity:
50
Catalog No. Quantity
6002871 50
1 options
Catalog No. 6002871 Supplier Solstice Supplier No. 9064
Medical-grade polypropylene with PE frits
  • All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar
Specifications
Quantity 50