Learn More
Solstice Polar Silica-Based SPE Columns, Cyanopropyl (CN), 200 mg/3 mL
Catalog No. 6002736
Change view
Click to view available options
Quantity:
50
Catalog No. Quantity
6002736 50
1 options
Catalog No. 6002736 Supplier Solstice Supplier No. 9062
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Medical-grade polypropylene with PE frits
  • All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar
Specifications
Quantity 50