Learn More
Solstice Polar Silica-Based SPE Columns, Aminopropyl (NH2), 100 mg/1 mL
Catalog No. 6002721
Change view
Click to view available options
Quantity:
100
Catalog No. Quantity
6002721 100
1 options
Catalog No. 6002721 Supplier Solstice Supplier No. HWL9070
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Medical-grade polypropylene with PE frits
  • All columns are normal phase and should be preconditioned with a non-polar solvent with the elution solvent being polar
Specifications
Quantity 100