missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pointe Scientific G6PD Reagents
Description
- Method: Glucose-6-Phosphate, Kinetic
- Format: Dry Powder
- Wavelength: 340nm
- Linearity: 21.0 U/g Hb
- Expected Values: 12.1+/-2.09 U/g Hb
- Reconstituted stability: 8 hours at RT, 5 days at 2° to 8°C
For the quantitative, kinetic determination of glucose-6-phosphate dehydrogenase (G6PD) in blood
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | G6PD Reagent Set |
| For Use With (Application) | Quantitative, kinetic determination of Glucose-6-Phosphate Dehydrogenase in Blood |
| Quantity | 10 x 6mL R1, 1 x 120mL R2 |
| Form | Dry Powder |