Learn More
Carson™ PocketMicro 20 to 60x LED Lighted Pocket Microscope with Aspheric Lens System DFS Item
LED lighted pocket microscope contains a 20 to 60x power zoom magnification range and internal aspheric lens system
Supplier:  Carson™ MM450
Catalog No. S81721
Only null left
Description
Description
  • Aspheric lens system allows light rays passing through lens to converge at a single focal point for a sharp, clear, distortion-free image
  • Great beginner microscope for kids, students, hobbyists and professionals
  • For examining plant buds, coins, stamps, bugs, nature, electronics, fabric, printing
  • Lightweight, compact design
  • Powered by 1 AA battery (not included)
  • Backed by Carson™'s limited lifetime warranty
  • For ideal use:
    • Hold microscope flat against surface and make sure LED light is activated
    • Begin at lowest 20x magnification and focus subject using focusing dial
    • Increase magnification to desired range using zoom dial
  • Warranty:
    • Backed by Carson™'s limited lifetime warranty
    • Carson™ will repair or replace pocket microscope if it is found to have manufacturing defects in materials or workmanship
    • Please contact Carson™ for additional warranty details
  • Specifications
    Specifications
    Field Microscope
    20X to 60X
    Product Suggestions
    Product Suggestions