Learn More
Pocket Loupes
Catalog No. S19027
Change view
Click to view available options
Magnification:
16x
Catalog No. Magnification
S19027 16x
1 options
Catalog No. S19027 Supplier Compass Industries Inc Supplier No. 794
Handy foldaway magnifier is always ready for close looks.
Have this compact pocket loupe always at hand for quick, close-up magnifying. The lens of the pocket loupe, available with 16X magnification, is protected by an integral fold-over metal case.
Specifications
Material Nickel-plated Brass Body, Glass Lens
Product Type Magnifier, Pocket Loupe
For Use With (Application) For quick, close-up magnifying
Magnification 16x
Includes Integral fold-over Metal case