missing translation for 'onlineSavingsMsg'
Learn More

Kova™ POC Urinalysis Dropper Controls

Catalog No. 22749800
Change view
Click to view available options
Quantity:
10 x 5 mL
4 x 25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22749800 10 x 5 mL
22749803 4 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22749800 Supplier Kova™ Supplier No. K88105

Urinalysis controls for smaller routine labs and point-of-care testing sites.

  • Designed for use in POC sites and smaller labs
  • Control values assigned for the most commonly used dipsticks and instruments
  • After opening, control is stabile for 30 days at room temp or until expiration when stored in refrigerator
  • Barcoded labels (lot/expiration)
  • Ready-to-use liquid controls in 2 levels (Normal/Abnormal)
  • 22 month shelf life from date of manufacture

Specifications

Certifications/Compliance CLSI and CLIA, ANSI Z400.5-2004
Color Yellow, green or amber
Content And Storage 2°C to 8°C
Detectable Analytes pH, Protein, Glucose, Ketones, Bilirubin, Hemoglobin, Nitrite, Urobilinogen, Specific Gravity, Leukocytes, Microalbumin, Creatinine, Ascorbic Acid
For Use With (Equipment) Urinalysis reagent strips
Intended Use Ready-to-use, liquid products intended for use as a control for qualitative and semi-quantitative procedures used in routine urinalysis
Quantity 10 x 5 mL
Form Liquid