missing translation for 'onlineSavingsMsg'
Learn More

Kova™ POC Urinalysis DipTube Controls with Microscopics

Catalog No. 22749801
Change view
Click to view available options
Format:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
22749801 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22749801 Supplier Kova™ Supplier No. K88115

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Urinalysis controls with microscopics for smaller routine labs and point-of-care testing sites.

  • Designed for use in POC sites and smaller labs
  • Control values assigned for the most commonly used dipsticks and instruments
  • After opening, control is stabile for 30 days at room temp or until expiration when stored in refrigerator
  • Barcoded labels
  • Contains microscopics
  • Ready-to-use liquid controls in 2 levels (Normal/Abnormal)
  • 22 month shelf life from date of manufacture

Specifications

Certifications/Compliance CLSI and CLIA, ANSI Z400.5-2004
Color Yellow, green or amber
Content And Storage 2°C to 8°C
Detectable Analytes pH, Protein, Glucose, Ketones, Bilirubin, Hemoglobin, Nitrite, Urobilinogen, Specific Gravity, Leukocytes, Microalbumin, Creatinine, Ascorbic Acid
Format Liquid
For Use With (Equipment) Urinalysis reagent strips
Intended Use Ready-to-use, liquid products intended for use as a control for qualitative and semi-quantitative procedures used in routine urinalysis
Quantity 6 x 15 mL
Form Liquid