Learn More
Lonza Walkersville Inc PNGM Primary Neuron Growth Medium BulletKit
Catalog No. NC9436348
Encompass
Clonetics PNGM Primary Neuron Growth Medium Kit (PNGM) was specifically developed for culturing embryonic primary rat and mouse neurons. The media kit is recommended for use with all of our primary rat and mouse neurons. Benefits:Serum-free mediumAllows for long-term maintenance and growth of neuronal cellsPromotes development of high quality neuronal networks without the need of an astrocyte feeder layerContent:1 x PNBM Basal Medium (CC-3256), 200 mL1 x PNGM Singlequots Growth Supplement Pack (CC-4462)
Catalog No. NC9436348 Supplier Lonza Walkersville Inc Supplier No. CC4461
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Clonetics PNGM Primary Neuron Growth Medium Kit (PNGM) was specifically developed for culturing embryonic primary rat and mouse neurons. The media kit is recommended for use with all of our primary rat and mouse neurons. Benefits:Serum-free mediumAllows for long-term maintenance and growth of neuronal cellsPromotes development of high quality neuronal networks without the need of an astrocyte feeder layerContent:1 x PNBM Basal Medium (CC-3256), 200 mL1 x PNGM Singlequots Growth Supplement Pack (CC-4462)