missing translation for 'onlineSavingsMsg'
Learn More

StatLab™ Plus Alcohol

Catalog No. STL70951
Change view
Click to view available options
Quantity:
4 x 1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STL70951 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STL70951 Supplier StatLab™ Supplier No. 70951

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, if in contact with skin, or if inhaled.

A custom isopropyl and methanol blend that can be used as a substitute for traditional ethanol blends.

  • 100% or 95% Plus alcohol (100% Plus alcohol with 5 parts deionized water)
  • Does not over-dehydrate small specimens
  • Great dehydrant for both small and large tissue specimens
  • May also be used in staining procedures

Specifications

Concentration 0.95
Quantity 4 x 1 gal.
Format Bottle