Learn More
Pluriselect pluriStrainer Mini (Cell Strainer) 20 um mesh-size, 500 pcs. (20x25), sterile
Supplier:  Pluriselect 431002070
Encompass
The pluriStrainer Mini is a sieving device (or cell strainer) for a wide range of laboratory applications where filtration and purification of liquids is required. It has been optimized for small sample volumes upto 700 ul. Its unique design allows fitting on a wide variation of tubes, e.g. 1.5 ml /2.0 ml reaction tubes, 15 ml conical centrifuge tubes, FACS-tubes, Cryo vials, 24 well plate and 48 well plate.
Catalog No. NC1730951