missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Pluronics Sacrificial Ink for Matribot™ Bioprinter

Catalog No. 01257119
Change view
Click to view available options
Quantity:
5 Syringes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01257119 5 Syringes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01257119 Supplier Corning™ Supplier No. 6157

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pluronics Sacrificial Ink is a ready-to-print sacrificial and support ink that exhibits excellent printability. Designed for use with Matribot™ Bioprinter.

  • Ready-to-print sacrificial and support ink that exhibits excellent printability
  • Easy to remove once printed by washing with cold PBS
  • Has been used for the fabrication of vascularized tissues, channels in microfluidic devices, and supporting scaffolds for complicated tissue constructs
  • Can also be diluted to 25% v/v with water to use as a learning tool to mimic dispensing of temperature-sensitive hydrogels
  • Designed for use with Corning™ Matribot™ Bioprinter
  • Specifications

    Color Transparent
    For Use With (Equipment) Corning Matribot Bioprinter
    Product Type Sacrificial Ink
    Capacity (Metric) 2.7 mL/Syringe
    Description Sterile-A Quality
    Model Pluronics
    For Use With (Application) Fabrication of vascularized tissues, channels in microfluidic devices, and supporting scaffolds for complicated tissue constructs
    Quantity 5 Syringes
    Storage Requirements 4°C to 8°C