Learn More
Biotium Pluronic F-127 20% Solution in DMSO
Catalog No. NC0322565
Encompass
Ready-to-use solution of Pluronic F-127, a nonionic detergent useful for solubilizing large dye molecules such as Fura-2/AM, Fluo-3/AM, Indo-1/AM and Rhod-2/AM to facilitate cell loading. Pluronic F-127 is a registered trade mark of BASF.
Catalog No. NC0322565 Supplier Biotium Supplier No. 59004
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ready-to-use solution of Pluronic F-127, a nonionic detergent useful for solubilizing large dye molecules such as Fura-2/AM, Fluo-3/AM, Indo-1/AM and Rhod-2/AM to facilitate cell loading. Pluronic F-127 is a registered trade mark of BASF.