Learn More
Molecular Probes™ Pluronic™ F-127, low UV absorbance
Description
Pluronic™ F-127, low UV absorbance
Pluronic F-127 is a nonionic, surfactant polyol (molecular weight approximately 12,500 daltons) that has been found to facilitate the solubilization of water-insoluble dyes and other materials in physiological media.1 Pluronic F-127 is commonly used to help disperse the acetoxymethyl (AM) esters of our ion indicators as well as our cell tracer dyes such as CFDA-SE.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 2 g |
For Research Use Only. Not for use in diagnostic procedures.