missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PluriSTEM™ C57BL/6N Murine Embryonic Stem Cells

Catalog No. SCC050
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC050 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC050 Supplier MilliporeSigma™ Supplier No. SCC050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as standard background for mouse genome engineering

  • Gender: Male
  • Karyotype: 40XY
  • Supplied frozen in basal medium supplemented with 50% FCS and 10% DMSO

Specifications

Content And Storage Place vials in liquid nitrogen immediately upon receipt until it is convenient to proceed to subculture.
Cell Type Murine Embryonic Stem Cells
For Use With (Application) Stem cell culture
Quantity 2 Vials

For Research Use Only