missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety Pleated Polypropylene Bouffant Cap

Catalog No. p-7240062
Change view
Click to view available options
Color:
Blue
Green
Orange
Red
White
Yellow
Size:
24 in.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19055101 White -
19055330 Orange -
19055331 Yellow -
19055332 Green -
19055333 Red -
19055335 Yellow -
19055336 Green -
19055337 Red -
296368083 Blue 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19055101 Supplier Keystone Safety Supplier No. 111NWI1021WHITE
Only null left

Comfortable, pleated bouffant caps feature an encapsulated elastic headband to keep hair in place. Available in a variety of sizes and colors.

  • Polypropylene
  • 100% Latex Free
  • Economical Protection
  • Sizes Available: 21, 24, 28 in
  • Flexible, pleated design that stretches over the worker's head
  • Disposable design for fresh, hygienic use every time

Specifications

Product Type Bouffant
Material Polypropylene
Color White
Disposable Single-use
Latex Free Yes
Model 111NWI-10
Sterility Non-sterile
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Style Pleated