missing translation for 'onlineSavingsMsg'
Learn More

International Enviroguard™ Pleated Bouffant Caps

Catalog No. p-7222261
Change view
Click to view available options
Color:
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19472783 White
19472786 -
19472784 White
19472787 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19472783 Supplier International Enviroguard™ Supplier No. S110NW21PL
Only null left

Pleated bouffant caps provide protection against dirt, grime, and particulates in non-hazardous environments. Available in blue or white, 21 in. or 24 in. diameter.

  • Provide a particulate barrier and helps prevent hair from contaminating a product or work environment
  • Low-linting, lightweight cover restrains hair and offers great ventilation
  • Designed to protect against dirt, grime, and particulates in non-hazardous environments
  • Pleated
  • Latex-free elastic
  • Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more

Specifications

Product Type Bouffant
Material Polypropylene
Color White
Disposable Single-use
Latex Free Yes
Model S110NW
For Use With (Application) Food Processing, Light Manufacturing and Construction, Environmental Cleanup
Style Pleated