Learn More
ACROBiosystems Inc ACROBiosystems Human PLAU / uPA Protein, His Tag (activated by trypsin)
Catalog No. 501956716
Encompass
Recombinant Protein;1MG;Human PLAU, His Tag (activated by trypsin) (PLU-H5228) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Leu 431 (Accession # P00749-1).;This protein carries a polyhistidine tag at the C-terminus. The active form of Human PLAU is a disulfide-linked heterodimer composed of long chain A (Ser 21 - Phe 177) and chain B (Ile 179 - Leu 431) with calculated MW of 17.8 kDa and 29.2 kDa. The long chain A is further cleaved to yield a short chain A (Lys 156 - Phe 177) and N-terminal fragment (Ser 21 - Lys 155) with calculated MW of 15.3 kDa. The protein migrates as 17 kDa (N-terminal fragment), 32-35 kDa (chain B) and 45-50 kDa (long chain A & chain B) under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.
Catalog No. 501956716 Supplier ACROBiosystems Inc Supplier No. PLUH52281MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;1MG;Human PLAU, His Tag (activated by trypsin) (PLU-H5228) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Leu 431 (Accession # P00749-1).;This protein carries a polyhistidine tag at the C-terminus. The active form of Human PLAU is a disulfide-linked heterodimer composed of long chain A (Ser 21 - Phe 177) and chain B (Ile 179 - Leu 431) with calculated MW of 17.8 kDa and 29.2 kDa. The long chain A is further cleaved to yield a short chain A (Lys 156 - Phe 177) and N-terminal fragment (Ser 21 - Lys 155) with calculated MW of 15.3 kDa. The protein migrates as 17 kDa (N-terminal fragment), 32-35 kDa (chain B) and 45-50 kDa (long chain A & chain B) under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.