missing translation for 'onlineSavingsMsg'
Learn More

Platinum wire, 0.6mm (0.024in) dia, hard, 99.95% (metals basis)

Catalog No. p-4408759
Change view
Click to view available options
Quantity:
10 cm
50 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43494BS 10 cm
AA43494BW 50 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43494BS Supplier Thermo Scientific Chemicals Supplier No. 043494BS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: approx. 6.06g/m, Temper: as drawn | CAS: 7440-06-4 | Pt | 195.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color White
Appearance Silvery white wire
Quantity 10 cm
Odor Odorless
Form Wire
Assay Percent Range 99.95% (Metals basis)
Chemical Name or Material Platinum
EINECSNumber 231-116-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only