missing translation for 'onlineSavingsMsg'
Learn More

Platinum wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis)

Platinum wire; 99.95% metal basis; Alfa Aesar; 0.5mm diameter; annelaled; 25cm length; 4.21g/m, Temper soft | CAS: 7440-06-4 | Pt | 195.08 g/mol

Supplier:  Thermo Scientific Chemicals 043288BY

 View more versions of this product

Catalog No. AA43288BY


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Platinum wire, Annealed
100 cm
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
6-4-7440
99.95% (metals basis)
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939

Chemical Identifiers

6-4-7440
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

RTECSNumber : TP2160000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only