missing translation for 'onlineSavingsMsg'
Learn More

Platinum wire, 0.3mm (0.012in) dia, 99.9% (metals basis)

Catalog No. p-6827159
Change view
Click to view available options
Quantity:
25 cm
100 cm
Catalog No. Quantity
AA43014BU 25 cm
AA43014BY 100 cm
2 options

Catalog No. AA43014BU

Supplier: Thermo Scientific Chemicals 043014BU

Only null left

Form: Approx.1.5g/m, 0.477g/ft. | CAS: 7440-06-4 | Pt | 195.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Appearance Silvery white wire
Density 21.45g/cm3 at 20°C
Quantity 25 cm
Odor Odorless
Form Wire
Assay Percent Range 99.9% (Metals basis)
Chemical Name or Material Platinum
EINECSNumber 231-116-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only