missing translation for 'onlineSavingsMsg'
Learn More

Platinum wire, 0.25mm (0.01in) dia, Premion™, 99.997% (metals basis)

Catalog No. p-5011349
Change view
Click to view available options
Quantity:
10 cm
250 cm
50 cm
Catalog No. Quantity
AA10958BS 10 cm
AA10958BW 50 cm
AA10958CB 250 cm
3 options

Catalog No. AA10958BS

Supplier: Thermo Scientific Chemicals 010958BS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-06-4 | Pt | 195.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Appearance Silvery white wire
Density 21.45g/cm3 at 20°C
Quantity 10 cm
Odor Odorless
Form Wire
Assay Percent Range 99.997% (Metals basis)
Chemical Name or Material Platinum
EINECSNumber 231-116-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only