missing translation for 'onlineSavingsMsg'
Learn More

Platinum wire, 0.0508mm (0.002in) dia, 99.95% (metals basis)

CAS: 7440-06-4 | Pt | 195.08 g/mol

$97.10 - $1549.93

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13479G1
View Documents
Thermo Scientific Chemicals
013479G1
1 m
Each for $97.10
Only null left
 
AA13479G5
View Documents
Thermo Scientific Chemicals
013479G5
5 m
Each for $391.90
Only null left
 
AA13479G9
View Documents
Thermo Scientific Chemicals
013479G9
25 m
Each for $1,549.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6-4-7440
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

6-4-7440
Odorless
Wire
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
99.95%
(metals basis)
1 m
≈0.043g/m
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
Platinum wire, 0.0508mm (0.002 in.) dia., Hard, Temper: as drawn
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only