missing translation for 'onlineSavingsMsg'
Learn More

Platinum Thinfoil, 0.004^+0.0025mm (0.00016^+0.0001in) thick, 99.95% (metals basis)

Catalog No. p-7049089
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Catalog No. Quantity
AA12868FF 25 x 25 mm
AA12868FI 50 x 50 mm
2 options

Catalog No. AA12868FF

Supplier: Thermo Scientific Chemicals 012868FF

Only null left

CAS: 7440-06-4 | Pt | 195.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Quantity 25 x 25 mm
Odor Odorless
Weight ≈0.05g/25x25mm
Form Foil
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Platinum Thinfoil, 0.004 (±)0.0025mm (0.00016 (±)0.0001 in.) thick, Not certified pinhole free
EINECSNumber 231-116-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only