missing translation for 'onlineSavingsMsg'
Learn More

Platinum Ruthenium foil, 0.05mm (0.002in) thick

Catalog No. AA42686FI
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42686FI 50 x 50 mm
AA42686FF 25 x 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42686FI Supplier Thermo Scientific Chemicals Supplier No. 042686FI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pt:Ru; 95.2:4.8

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Color Silver to Gray
Chemical Name or Material Platinum Ruthenium
Appearance Silver-gary foil
Quantity 50 x 50 mm
Odor Odorless
Form Foil
Molecular Formula Pt:Ru; 95.2:4.8
MDL Number MFCD00798552

RUO – Research Use Only