missing translation for 'onlineSavingsMsg'
Learn More

Platinum powder, -22 mesh, Premion™, 99.99% (metals basis)

CAS: 7440-06-4 | Pt | 195.08 g/mol

$183.59 - $2704.45

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1067201
View Documents
Thermo Scientific Chemicals
01067201
0.25 g
Each for $183.59
Only null left
 
AA1067203
View Documents
Thermo Scientific Chemicals
01067203
1 g
Each for $582.55
Only null left
 
AA1067206
View Documents
Thermo Scientific Chemicals
01067206
5 g
Each for $2,704.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6-4-7440
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

6-4-7440
Powder
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
Premion
Platinum powder
Silver-White
0.25 g
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
99.99%
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 231-116-1

RTECSNumber : TP2160000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only