missing translation for 'onlineSavingsMsg'
Learn More

Platinum powder, -200+400 mesh, 99.98% (metals basis)

Catalog No. p-7080154
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4683003 1 g
AA4683006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4683003 Supplier Thermo Scientific Chemicals Supplier No. 04683003
Only null left

CAS: 7440-06-4 | Pt | 195.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Physical Form Powder
Quantity 1 g
Percent Purity 99.98%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Platinum powder
Hazard Category H228
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 231-116-1
RTECSNumber TP2160000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only