missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) oxide, 83% Pt

CAS: 1314-15-4 | O2Pt | 227.08 g/mol

$386.71 - $1333.10

Chemical Identifiers

CAS 1314-15-4
Molecular Formula O2Pt
Molecular Weight (g/mol) 227.08
MDL Number MFCD00011184
InChI Key YKIOKAURTKXMSB-UHFFFAOYSA-N
Synonym Adams' catalyst
IUPAC Name dioxoplatinum
SMILES O=[Pt]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC195320010
SDS View Documents
Thermo Scientific Chemicals
195320010
1 g Glass bottle
Each for $386.71
Only null left
 
AC195320050
SDS View Documents
Thermo Scientific Chemicals
195320050
5 g Glass bottle
Each for $1,333.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1314-15-4
227.08
YKIOKAURTKXMSB-UHFFFAOYSA-N
dioxoplatinum
O2Pt
MFCD00011184
Adams' catalyst
O=[Pt]=O

Specifications

450.0°C
Brown
1 g
PtO2
1479
Adams' catalyst
YKIOKAURTKXMSB-UHFFFAOYSA-N
dioxoplatinum
227.09
Glass bottle
1314-15-4
Powder
O2Pt
MFCD00011184
15, 7642
Solubility in water: insoluble
O=[Pt]=O
227.08
83% (Pt)
Platinum(IV) oxide, 83% Pt
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause fire or explosion; strong oxidizer.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

EINECSNumber : 215-223-0

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only